BACEA-1010
A-1010.0025 A-1010.0005
A-1010.0005BAEA 161.18 USD
A-1010.0005BA A-1010.0025BA
BACEA-1010
Sequence: Boc-His(Boc)-OH benzene solvate
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.