Would you like to log in again?
Stablcal® Turbidity Standard, 0.30 NTU, 100mL, Hach
Supplier: Hach
Stablcal®
26979-42 2697942
97026-426EA 65.74 USD InStock
CA26979-42 97026-426
Stablcal® Turbidity Standard, 0.30 NTU, 100mL, Hach
Standards Turbidity Standards
Stablcal® Formazin Turbidity Standard, 0.30 NTU.
  • Precise, Premixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty