Phenol 2 Reagent Powder Pillows, Hach
Supplier: Hach
183699 183699
97026-610PK 115.57 USD
97026-610 CA1836-99
Phenol 2 Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of phenols by the 4-Aminoantipyrine Method (Hach method 8047).
Ordering information: Pack of 100 powder pillows.