Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Amino Acid F Dilution Solvent, for Rapid Liquid Silica, 475 mL, Hach
Supplier: Hach
2353011 2353011
89231-046PK 36.44 USD InStock
CA11017-074 89231-046
Amino Acid F Dilution Solvent, for Rapid Liquid Silica, 475 mL, Hach
Environmental Test Kits
For determination of Ultra Low Range silica by the Heteropoly Blue Rapid Liquid method
Hach method 8282 Range: 3 - 1000 μg/L SiO2. Used with Amino Acid F Reagent Powder (2651155). Rapid Liquid™ methods offer an economical way to analyze multiple samples and to measure trace concentrations. Requires Pour-Thru Cell.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty