Total Inorganic Nitrogen Pretreatment Reagent Set, Hach
Supplier: Hach
2604945 2604945
89231-418EA 158.3 USD InStock
CA11024-470 89231-418
Total Inorganic Nitrogen Pretreatment Reagent Set, Hach
Environmental Test Kits
Pretreatment Reagent Set for determination of Total Inorganic Nitrogen by the Titanium Trichloride Reduction Test 'N Tube™ Method
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Used with Nitrogen-Ammonia Reagent Set, TNT, AmVer (Salicylate), LR (2604545). 25-50 tests (dependent upon the number of reagent blank tests performed).
Hach Method 10021.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.