ORP110-GS Standard ORP Probe, Hach
Supplier: Hach
ORP110-GS ORP110-GS
82020-480EA 271.24 USD
CA11026-970 82020-480
ORP110-GS Standard ORP Probe, Hach
ORP110-GS Standard ORP Probe with BNC Connector, 3 ft
cable, 12 x 110 mm body, and Platinum band sensor.
Ordering information: 6 month warranty.