High-Speed Mini Centrifuge Microspin 12, Grant Instruments
MICROSPIN12USA
89402-942EA 1958.49 USD InStock
89402-942
High-Speed Mini Centrifuge Microspin 12, Grant Instruments
Centrifuges Benchtop Centrifuges
This compact, high-speed benchtop mini centrifuge features a built-in rotor for 12x1.5/2mL microtubes.
  • Centrifugation speed: 100-14,500rpm, RCF 12,400 x g
  • Fixed angular aluminium rotor accommodates 12 x 1.5/2ml microtubes, supplied with adapters for 0.2ml and 0.5ml tubes
  • Timed operation (1 to 30 minutes), with automatic switch-off
  • Suitable to use in cold rooms, operating temperature ambient +4°C to +25°C
  • Very small footprint
Certifications: CE marked.
Ordering information: It is ideal for biomedical laboratories. Units are supplied with a manufacturer's two-year warranty. Additional rotors are available.