Multi-Spin™ PCV-6000, Grant Instruments
PCV-6000USA
89402-912EA 316.82 USD
89402-912
Multi-Spin™ PCV-6000, Grant Instruments
Highly versatile and efficient variable-speed combined centrifuge/vortex mixer
  • Centrifugation speed: Up to 6000rpm
  • Vortex mixing modes - soft, medium and hard with regulated timer of 1 to 20 seconds, with automatic switch-off
  • Choice of interchangeable rotors for different microtube sizes combinations and for 0.2ml strips
  • Can be used at temperatures from +4ºC to +40ºC
Spin-mix-spin technology can save considerable time by automatically performing a cycling program of sample mixing and spinning 12 tubes at once, when compared with removing the tubes for vortexing after every spin.
Programmed centrifugation and mix operations or independent centrifuging and vortex-mixing of multiple microtubes and 0.2ml strips
Certifications: CE marked.
Ordering information: Units are supplied with a manufacturer's two-year warranty. Additional rotors are available.