sensION Sodium Platinum Series Combination Ion Selective Electrode (ISE), Hach
Supplier: Hach
5192500 5192500
97021-598EA 1059.88 USD InStock
CA11014-078 97021-598
sensION Sodium Platinum Series Combination Ion Selective Electrode (ISE), Hach
Electrodes ISE Electrodes
Sodium Platinum Series Combination Ion Selective Electrode (ISE) with BNC connector.
  • Accurate
  • Easy to Use
  • Rugged and Durable
  • Flexible
Electrode includes two electrolyte cartridges and a manual. Compatible with sensION and H-Series systems.