Tweezers, Angulated Wire Cutting, Excelta
Supplier: EXCELTA CORPORATION
14A-GW 15A-GW-M 15A-GW-OF 15A-GW-PB 15A-RW 15A-RW-M 15A-HT 15A-RW-OF 15A-RW-S 15A-GW-S 15A-GW-ET
89411-882EA 394.93 USD InStock
89411-882 89411-914 89411-920 89411-936 89411-924 89411-926 89411-950 89411-930 89411-940 89411-938 89411-922
Tweezers, Angulated Wire Cutting, Excelta
Forceps
Cutting tweezers are made from high grade carbon steel and feature angulated cutting heads to get into tight places
They offer a clean and precise flush cut on soft wire.