sensION Refillable Combination pH Electrode, BNC and 3.5 mm Phone Connectors, Hach
Supplier: Hach
5194011
97021-604EA 528.76 USD
97021-604
sensION Refillable Combination pH Electrode, BNC and 3.5 mm Phone Connectors, Hach
Electrodes pH Electrodes
Standard combination pH probe with a refillable reference and built-in temperature sensor
Contains fixed cable with BNC and 3.5 mm phone connectors compatible Hach One series meters.