Would you like to log in again?
Stablcal® Turbidity Standards Calibration Kit, 2100AN/AN IS Turbidimeter
Supplier: Hach
Stablcal®
2659500
97026-810EA 585.87 USD InStock
97026-810
Stablcal® Turbidity Standards Calibration Kit, 2100AN/AN IS Turbidimeter
Standards Turbidity Standards
Stablcal® Standards Calibration Kit, for 2100AN and 2100ANis Laboratory Turbidimeters
  • Precise and pre-mixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability
  • Saves time and labor
  • Confidence in results
Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter.
Ordering information: Set of 6. Kit includes 500 mL bottles of <0.1, 20, 200, 1000, 4000 NTU standard and a sealed vial of 7500 NTU standard.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty