Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Stablcal® Turbidity Standard, 800 NTU, Hach
Supplier: Hach
Stablcal® StablCal®
2660549 2660549 2660542 2660542
97026-476EA 115.99 USD InStock
CA11016-412 97026-476 CA11016-418 97026-398
Stablcal® Turbidity Standard, 800 NTU, Hach
Standards Turbidity Standards
Stablcal® Turbidity Standard for all Turbidimeters.
  • Precise, Premixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: 800.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty