Ammonium sulfate, cartridge Reference electrolyte

Synonyms: Diammonium sulfateSulphuric acid diammonium saltDiammonium sulphateSulfuric acid diammonium salt

2597102
97021-768PK 25.96 USD
97021-768
Ammonium sulfate, cartridge Reference electrolyte
Ammonium sulphate

Ammonium Sulfate Reference Cartridge


Reference electrolyte for nitrate determination by ISE.


Ordering information: Pack of 2.

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR