Bromophenol blue solution acid-base indicator
Supplier: Hach
Synonyms:
Bromphenol Blue Sultone Form
, Tetrabromophenol-sulfonephthalein
, 3,3',5,5'-Tetrabromophenolsulfonphthalein
, Tetrabromophenol-sulphonephthalein
, BPB
, 3,3',5,5'-Tetrabromophenolsulphonphthalein
pH indicator for acidity and alkalinity titrations
Visual transition interval: pH 3.0 (yellow)-pH 4.6 (blue).
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Formula:
C₁₉H₁₀Br₄O₅S MW: 669.97 g/mol |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...