Tannin-Lignin Reagent Set, Hach
Supplier: Hach
22446-00 2244600
89231-348PK 128.49 USD InStock
CA22446-00 89231-348
Tannin-Lignin Reagent Set, Hach
Environmental Test Kits
For determination of Tannin and Lignin by the Tyrosine Method
Hach Method 8193. Range: 0.1 - 9.0 mg/L. Sample Size: 25 mL. Set Includes: Sodium Carbonate Solution and TanniVer™ 3 Tannin-Lignin Reagent for up to 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.