Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Ionic strength adjustor (ISA) Buffer solution, for use with Nitrate Ion Selective Electrode
Supplier: Hach
2488349
97021-528EA 47.54 USD InStock
97021-528
Ionic strength adjustor (ISA) Buffer solution, for use with Nitrate Ion Selective Electrode
Ionic strength adjustor (ISA)
Nitrate Ionic Strength Adjustor (ISA) Buffer Solution for sample treatment in nitrate determination by ion selective electrode.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty