Detergents Reagent Set, Hach
Supplier: Hach
2446800 2446800
89231-396EA 302.43 USD
CA11024-846 89231-396
Detergents Reagent Set, Hach
Environmental Test Kits
For determination of Detergents (Surfactants) by the Crystal Violet method (Hach Method 8028)
Range: 0.002 - 0.275 mg/L LAS. Reagent Set includes: Benzene, Buffer Solution and Detergent Reagent Powder Pillows for 70 - 100 tests. Note: No Air Shipments - see Product Number 2446801 for Air Freight Configuration.
Ordering information: No Air Shipments.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.