Multi-Type Calibrator Thermometer with NIST Certificate, Extech
433201-NIST
89087-408EA 681.47 USD
89087-408
Multi-Type Calibrator Thermometer with NIST Certificate, Extech
The Multi-Type Calibrator Thermometer with the ability to source and measure 8 thermocouple type devices J, K, T, E, C, R, S, and N
CE marked. Includes NIST certificate.
High Accuracy Calibration function simulates precision thermocouple outputs for use in calibrating thermometers, transmitters,controllers or recorders. The microprocessor assures maximum accuracy to 0.15% of reading over wide ranges.
Ordering information: One-year warranty. Complete with universal AC adaptor and AU/EU/UK/US plugs.