General Purpose Laboratory Incubators, SHEL LAB
SLM722 SMI12 SMI6 SMI2 SMI11
89409-318EA 4422.6 USD
89409-318 89409-322 89409-316 89409-314 89409-320
General Purpose Laboratory Incubators, SHEL LAB
SHEL LAB General Purpose Incubators are the ideal solution for industrial protocols, biological research and environmental studies that demand accurate and repeatable results
Heated doors and a unique air jacket design achieve precise temperature uniformity. An independent secondary temperature controller offers the added safety and security of over temperature production.
The GI series models include a sealed, inner glass door which provides a view into the chamber without compromising samples or the chamber environment. Stainless steel panels and doors reduce contamination, provide durability and allow for easy cleaning. One internal electrical outlet. One access port.
Their best in class temperature uniformity is usually found only in more expensive, application specific incubators. SHEL LAB General Purpose Incubator's wide temperature and size range make them a perfect solution for any lab.
Certifications: TUV, UL, CSA, EU, CE.
Ordering information: Two-year limited warranty.