Would you like to log in again?
µLPlate® Component Kits, with Red Caps, MicroLiter
Supplier: MicroLiter Analytical Supply
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials. The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer secure vessels that maintain even the most volatile samples.
Polypropylene μLplates™ have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-Well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options lets users choose the specifications appropriate for any application.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Material Well Volume Color Septa Type Well Bottom Shape Supplier No. VWR Catalog Number Unit Price Quantity
Glass Insert and Red Cap Polypropylene 1.0 mL Clear Precut FEP/Silicone Conical 07-0020R1000 97044-634 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 350 µL Clear Precut FEP/Silicone Conical 07-0020R-201 97044-644 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 150 µL Clear Precut FEP/Butyl Conical 07-0021R-101 97044-718 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Clear PTFE/Silicone Stopper Conical 07-0045R-300 97044-822 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 350 µL Clear Precut PTFE/Silicone Conical 07-0075R-201 97044-910 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Clear PTFE/Silicone Conical 07-0070R-300 89231-776 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 150 µL Clear PTFE/Silicone Conical 07-0070R-101 89231-766 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 150 µL Clear Precut FEP/Silicone Conical 07-0020R-101 97044-638 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Clear Precut FEP/Silicone Conical 07-0020R-300 97044-648 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 700 µL Amber Precut FEP/Butyl Round 07-0021R1202 97044-722 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Amber PTFE/Silicone Stopper Conical 07-0045R1002 97044-810 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.2 mL Clear Precut PTFE/Silicone Round 07-0075R1200 97044-906 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Amber PTFE/Silicone Conical 07-0070R1002 89231-764 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 700 µL Amber Precut FEP/Silicone Round 07-0020R1202 97044-642 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Amber Precut FEP/Butyl Conical 07-0021R1002 97044-716 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 2.2 mL Clear Precut FEP/Butyl Round 07-0021R2200 97044-726 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.2 mL Clear PTFE/Silicone Stopper Round 07-0045R1200 97044-814 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.0 mL Clear Precut PTFE/Silicone Conical 07-0075R1000 97044-900 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.2 mL Clear PTFE/Silicone Round 07-0070R1200 89231-768 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 700 µL Amber PTFE/Silicone Stopper Round 07-0045R1202 97044-816 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 2.2 mL Clear PTFE/Silicone Stopper Round 07-0045R2200 97044-820 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 2.2 mL Clear Precut PTFE/Silicone Round 07-0075R2200 97044-912 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 700 µL Amber PTFE/Silicone Round 07-0070R1202 89231-770 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.2 mL Clear Precut FEP/Silicone Round 07-0020R1200 97044-640 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.0 mL Clear Precut FEP/Butyl Conical 07-0021R1000 97044-714 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 350 µL Clear Precut FEP/Butyl Conical 07-0021R-201 97044-724 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Clear Precut FEP/Butyl Conical 07-0021R-300 97044-728 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 150 µL Clear PTFE/Silicone Stopper Conical 07-0045R-101 97044-812 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Amber Precut PTFE/Silicone Conical 07-0075R1002 97044-902 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 700 µL Amber Precut PTFE/Silicone Round 07-0075R1202 97044-908 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 350 µL Clear PTFE/Silicone Conical 07-0070R-201 89231-772 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.0 mL Clear PTFE/ Silicone Conical 07-0070R1000 89231-762 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Amber Precut FEP/Silicone Conical 07-0020R1002 97044-636 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 2.2 mL Clear Precut FEP/Silicone Round 07-0020R2200 97044-646 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.2 mL Clear Precut FEP/Butyl Round 07-0021R1200 97044-720 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 1.0 mL Clear PTFE/Silicone Stopper Conical 07-0045R1000 97044-808 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 350 µL Clear PTFE/Silicone Stopper Conical 07-0045R-201 97044-818 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 150 µL Clear Precut PTFE/Silicone Conical 07-0075R-101 97044-904 Pack of 100 Retrieving
Small Business Enterprise
Glass Insert and Red Cap Polypropylene 500 µL Clear Precut PTFE/Silicone Conical 07-0075R-300 97044-914 Pack of 100 Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty