Would you like to log in again?
µLPlate® with Inserts Installed and Red CapMat, MicroLiter
Supplier: MicroLiter Analytical Supply
A comprehensive line of 96-well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials. The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer you secure vessels to maintain even the most volatile samples.
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. You may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options let you choose the specifications appropriate for your application.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Material Well Volume Color Septa Type Well Bottom Shape Supplier No. VWR Catalog Number Unit Price Quantity
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Clear PTFE/Silicone Conical 07-3070MR300 89231-838 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 150 µL Clear Precut FEP/Butyl Conical 07-1221MR101 97044-992 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 350 µL Clear PTFE/ Silicone Stopper Conical 07-2045MR201 97045-132 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Clear Precut PTFE/Silicone Conical 07-3075MR300 97045-292 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 350 µL Clear Precut FEP/Silicone Conical 07-2020MR201 97045-082 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.2 mL Clear Precut FEP/Butyl Round 078021MR1200 97045-578 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Clear Precut FEP/Butyl Conical 07-3021MR300 97045-236 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 2.2 mL Clear Precut FEP/Silicone Round 078020MR2200 97045-462 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.2 mL Clear PTFE/ Silicone Stopper Round 078045MR1200 97045-698 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.2 mL Clear Precut PTFE/Silicone Round 078075MR1200 97045-822 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 700 µL Amber PTFE/Silicone Round 078070MR1202 89231-924 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 150 µL Clear PTFE/Silicone Conical 07-1270MR101 89231-790 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 700 µL Amber Precut FEP/Butyl Round 078021MR1202 97045-580 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 700 µL Amber PTFE/ Silicone Stopper Round 078045MR1202 97045-700 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 700 µL Amber Precut PTFE/Silicone Round 078075MR1202 97045-824 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 150 µL Clear Precut FEP/Silicone Conical 07-1220MR101 97044-968 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 350 µL Clear Precut FEP/Butyl Conical 07-2021MR201 97045-106 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Clear PTFE/ Silicone Stopper Conical 07-3045MR300 97045-262 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 700 µL Amber Precut FEP/Silicone Round 078020MR1202 97045-460 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.0 mL Clear PTFE/Silicone Conical 078070MR1000 89231-918 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.0 mL Clear Precut FEP/Butyl Conical 078021MR1000 97045-574 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.0 mL Clear PTFE/ Silicone Stopper Conical 078045MR1000 97045-694 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.0 mL Clear Precut PTFE/Silicone Conical 078075MR1000 97045-818 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 350 µL Clear Precut PTFE/Silicone Conical 07-2075MR201 97045-162 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.0 mL Clear Precut FEP/Silicone Conical 078020MR1000 97045-454 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.2 mL Clear Precut FEP/Silicone Round 078020MR1200 97045-458 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 2.2 mL Clear PTFE/Silicone Round 078070MR2200 89231-926 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Amber Precut PTFE/Silicone Conical 078075MR1002 97045-820 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Amber PTFE/Silicone Conical 078070MR1002 89231-920 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 350 µL Clear PTFE/Silicone Conical 07-2070MR201 89231-814 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 150 µL Clear Precut PTFE/Silicone Conical 07-1275MR101 97045-046 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Clear Precut FEP/Silicone Conical 07-3020MR300 97045-212 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Amber Precut FEP/Silicone Conical 078020MR1002 97045-456 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Amber Precut FEP/Butyl Conical 078021MR1002 97045-576 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 500 µL Amber PTFE/ Silicone Stopper Conical 078045MR1002 97045-696 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 1.2 mL Clear PTFE/Silicone Round 078070MR1200 89231-922 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 2.2 mL Clear Precut FEP/Butyl Round 078021MR2200 97045-582 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 2.2 mL Clear PTFE/ Silicone Stopper Round 078045MR2200 97045-702 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 2.2 mL Clear Precut PTFE/Silicone Round 078075MR2200 97045-826 Each Retrieving
Small Business Enterprise
Plate with Glass Inserts and Red CapMat Polypropylene 150 µL Clear PTFE/ Silicone Stopper Conical 07-1245MR101 97045-016 Each Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty