Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Falcon® 384-Well Small Volume Tissue Culture Treated Microplates, Corning®
Supplier: Corning
Falcon™
353380 353379 353379 353380
89218-116CS 973.42 USD InStock
89218-116 CA89218-114 89218-114 CA89218-116
Falcon® 384-Well Small Volume Tissue Culture Treated Microplates, Corning®
Microplates Cell Culture Plates
The tissue culture treated surface is permanently hydrophilic
It supports a wide range of applications and many important cell types, including primary, stem, neuronal, mesenchymal, hepatocyte and endothelial cells. All plates are supplied sterile, without lids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty