Falcon® 1536-Well Tissue Culture Treated Microplates, Corning®
Falcon™
353386 353385 353383 353383 353381 353381 353386 353384 353385 353382 353382 353384
89218-112CS 2016.58 USD
89218-112 CA89218-104 CA89218-106 89218-106 CA89218-108 89218-108 CA89218-112 CA89218-110 89218-104 CA89218-102 89218-102 89218-110
Falcon® 1536-Well Tissue Culture Treated Microplates, Corning®
Microplates Cell Culture Plates
The tissue culture treated surface is permanently hydrophilic
It supports a wide range of applications and many important cell types, including primary, stem, neuronal, mesenchymal, hepatocyte and endothelial cells. All plates are supplied sterile, without lids.