Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Falcon® 96-Well Tissue Culture Treated Microplates, Corning®
Supplier: Corning
Falcon™
353376 353376 353377 353377 353219 353219
89218-096CS 247.45 USD InStock
CA11008-408 89218-096 89218-098 CA11008-410 89022-352 CABD353219
Falcon® 96-Well Tissue Culture Treated Microplates, Corning®
Microplates Cell Culture Plates
The tissue culture treated surface is permanently hydrophilic
It supports a wide range of applications and many important cell types, including primary, stem, neuronal, mesenchymal, hepatocyte and endothelial cells. All plates are supplied sterile, with lids.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty