Potassium 2 Reagent Solution Pillows, Hach
Supplier: Hach
1432298 14322-98
89230-966PK 20.47 USD InStock
89230-966 CA14322-98
Potassium 2 Reagent Solution Pillows, Hach
Environmental Test Kits
For potassium determination in water, soil and plant tissues by the tetraphenylborate method
Hach Method 8049 Sample size: 25 mL.
Ordering information: Pack of 25 powder pillows.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.