sensION Gel Filled Combination pH Electrode, BNC and DIN Connectors, Hach
Supplier: Hach
5193522 51935-22
97021-584EA 626.73 USD
97021-584 CA51935-22
sensION Gel Filled Combination pH Electrode, BNC and DIN Connectors, Hach
Electrodes pH Electrodes
Standard combination pH probe with a non-refillable gel reference and built-in temperature sensor
Contains fixed cable with BNC and DIN connectors compatible Hach EC series meters.