Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Nitrogen-Nitrate Standard Solution, 10 mg/L as NO₃-N (NIST), 500 mL, Hach
Supplier: Hach
30749
97021-478EA 27.4 USD InStock
97021-478 CA307-49
Nitrogen-Nitrate Standard Solution, 10 mg/L as NO₃-N (NIST), 500 mL, Hach
Standards Environmental Analysis Standards
For accuracy checks and instrument calibrations in nitrate nitrogen determinations.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty