Pocket Colorimeter™ II, Lead Test (LeadTrak®), Hach
5870021
97026-914EA 804.67 USD InStock
97026-914
Pocket Colorimeter™ II, Lead Test (LeadTrak®), Hach
The new Pocket Colorimeter™ II Filter Photometer is a true go-anywhere instrument.
  • Single Parameter go anywhere portable photometer
  • USEPA approved for drinking water and wastewater
  • FactoryProgrammed
  • Waterproof, Lightweight, Rugged
  • Backlit LCD Display, Data Logging, Recall
It's lightweight and battery operated, suitable for extended field work or quick, on-the-spot process monitoring
What's in the Box - Pocket Colorimeter II Analysis System using Lead (LeadTrak™) Fast Column Extraction Method. Testing range: 5 to 150 µg/L as Pb. Use for screening first-draw samples for high concentrations of lead in drinking water; helps meet compliance with Lead/Copper Rule. Includes Lead Pocket Colorimeter II, reagents (20 tests), standard, manual and carrying case. Each Pocket Colorimeter II Instrument comes complete as a ready-to-use kit that includes a reagent set (pre-measured unit dose reagents), sturdy custom carrying case, and manual.
The instrument has two channels in which measurements can be made. Each channel will accept a user-defined calibration curve. Up to 10 standards can be used to determine the calibration. The curve is generated by a point-to-point straight line segment between each standard used. Linear and non-linear positive or negative slope calibrations can be performed. A calibration curve may also be manually entered from the keypad if a previously determined curve has been made on the Colorimeter. At least two data pairs (concentration and absorbance) are required.
Ordering information: 2 yr warranty.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.