Would you like to log in again?
DPD Total Chlorine Reagent Powder Pillows, 5 mL, Hach
Supplier: Hach
21982-46 2198246
89231-334PK 14.1 USD InStock
CA21982-46 89231-334
DPD Total Chlorine Reagent Powder Pillows, 5 mL, Hach
Environmental Test Kits
For determination of total chlorine with Hach's Chlorine Visual Test Kits
Sample Size: 5 mL. Range: up to 3.5 mg/L Cl2. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Ordering information: Pack of 50 Powder Pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty