Would you like to log in again?
Nitrogen-Nitrate Standard Solution, 1000 mg/L as NO3-N (NIST), 500 mL, Hach
Supplier: Hach
1279249 1279249
97021-442EA 41.62 USD InStock
CA97021-442 97021-442
Nitrogen-Nitrate Standard Solution, 1000 mg/L as NO3-N (NIST), 500 mL, Hach
Standards Environmental Analysis Standards
For accuracy checks in nitrate determinations
1000 mg/L as NO3-N; 4430 mg/L as NO3-
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty