Manganous Sulfate Powder Pillows, Hach
Supplier: Hach
107166 1071-66
97026-680PK 23.44 USD
97026-680 CA1071-66
Manganous Sulfate Powder Pillows, Hach
Environmental Test Kits
For determination of Dissolved Oxygen by the Winkler titration method
Range: 1 to >10 mg/L. Hach methods 8215 and 8229. Sample size: 300 mL.
Ordering information: 50/pk.