MEL/P-A Safe Drinking Water Lab, Hach
Supplier: Hach
2569600
89231-134EA 3670.21 USD
89231-134
MEL/P-A Safe Drinking Water Lab, Hach
Environmental Test Kits
Economical portable lab monitors key water quality parameters plus Total Coliforms and E. Coli (Presence-Absence Method).
  • Chemistry and sampling/filtration consumables only. Choose media and order separately
  • Nitrate and Chlorine determined using visual color discs
  • Portable incubator used to determine total and fecal coliforms directly in the field
  • Measure pH and TDS with portable testers
Contains portable incubator and rack, UV lamp, Whirlpak bags w/thiosulfate, Pocket Pal pH and TDS testers, thermometer, permanent marker, germicidal cloths, test kit components and reagents (100 tests each) for Free and Total Chlorine and Nitrate, and pH buffers.
Contains mercury. Dispose of according to local, state and federal regulations.