Grant QBA Analogue Block Heater, Grant Instruments
QBA1L QBA2L
97011-880EA 767.87 USD InStock
97011-880 97011-882
Grant QBA Analogue Block Heater, Grant Instruments
Block Heaters Analog Block Heaters
The QBA series of Analogue Block Heaters presents a contemporary, high-quality solution at a cost-effective price for heating applications up to 100°C.
Designed for durability and ease of use, the QBA series consists of two models with one or two interchangeable blocks. A comprehensive range of accessory blocks for tubes of different diameters ensures maximum flexibility for a variety of applications. The two-block system, QBA2, also provides the option of blocks for accommodating standard microplates and microtube strips.
Robustly constructed in streamlined coolwall aluminium and chemical-resistant plastic, the units have a small footprint and low profile to optimize bench space. A sloping fascia is complemented with an easy-to-use analogue interface. This, combined with a precision thermostat allowing a 2°C setting resolution, increases usability.
The Grant QBA Analogue Block Heaters feature ambient +5 to 100°C operation, with a setting resolution of 2°C. They offer temperature stability of ± 1.0°C, and temperature uniformity of ± 1.0°C at 37°C. Users are offered the choice of one or two block systems, with a wide range of accessory blocks for varied applications.
Certifications: CE approved.
Ordering information: Grant QBA Analogue Block Heaters are supplied with a block extraction tool for easy and safe removal of blocks. All units also include a manufacturer's three-year warranty. Interchangeable blocks must be ordered separately. A 230V version is available. Contact your VWR representative for additional information.