Cobalt/Nickel Reagent Set, PAN Method, 10 mL, Hach
Supplier: Hach
2651600 2651600
97026-266EA 184 USD
CA97026-266 97026-266
Cobalt/Nickel Reagent Set, PAN Method, 10 mL, Hach
Environmental Test Kits
For determination of Cobalt and Nickel by the PAN method
Hach Method 8078 (Co) and 8150 (Ni). Range: 0.01 - 2.00 mg/L Co, 0.006 - 1.000 mg/L Ni. Sample Size: 10 mL. Reagent set includes EDTA Reagent Powder Pillows, Phthalate-Phosphate Reagent Powder Pillows, and PAN Indicator Solution (0.3%) for approximately 100 tests.