Would you like to log in again?
SDL100 pH/ORP/Temperature Datalogger, Extech
Supplier: Extech
SDL100 SDL100-NIST
89370-232EA 130.22 USD InStock
89370-232 10171-834
SDL100 pH/ORP/Temperature Datalogger, Extech
Electrochemical Meters pH and Multi-Parameter Meters
pH/ORP/Temperature Datalogger that records data on an SD card in Excel® format
Complete with 6 AA batteries, SD card, sample buffers, mini pH electrode (60120B), temperature probe (850188), and hard carrying case.
The SDL100 features a Large backlit dual LCD screen with dual display of pH or mV and Temperature. It stores 99 readings manually and 20M readings via 2G SD card with a user programmable sampling rate. 3 point calibration ensures the best linearity and accuracy. Other features include Min/Max and Data Hold functions, and Auto power off functions.
Certifications: CE marked.
Ordering information: Three year warranty (meter); 6 months conditional (electrode).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty