ExStik®II Conductivity Meter, Extech
Supplier: Extech
EC500
89030-394EA 244.41 USD
89030-394
ExStik®II Conductivity Meter, Extech
Electrochemical Meters Conductivity Meters
Combination flat surface pH electrode with autoranging high accuracy Conductivity cell
The EC500 Measures 5 parameters including Conductivity, TDS, Salinity, pH, and Temperature using one electrode. It features memory that stores up to 25 labeled readings and a RENEW feature alerts user when electrode needs replacement.
Certifications: CE marked. IP57.
Ordering information: One year warranty (meter); 6 months conditional (electrode). Complete with combination pH/Conductivity/TDS cell, electrode, sensor cap, sample cup with cap, four 3V CR2032 batteries, and 48" neckstrap. Order Conductivity standards and pH buffers separately.