Direct reading of Total Chlorine down to 0.01 ppm.
- Low detection limit (down to 0.01 ppm)
- Unaffected by sample color or turbidity
- Memory stores, tags and recalls up to 15 readings
Provides a direct reading of Total Chlorine that provides fast and easy measurements (less than 2 minutes). US EPA approved as an acceptable method for wastewater compliance monitoring of Total Chlorine.
Certifications: CE marked. IP57.
Ordering information: One year warranty (meter); 6 months conditional (electrode). Complete with meter and flat surface Chlorine electrode, ExTab™ reagent tablets, protective sensor cap, somaple cup with cap, four 3V button batteries, and 48" neckstrap.