Would you like to log in again?
Falcon® Untreated Cell Culture Plates, Corning®
Supplier: Corning
351178 351143 351143 351146 351178 351146
15705-059CS 203.98 USD InStock
CA15705-062 CA15705-058 15705-059 CA15705-056 15705-062 15705-056
Falcon® Untreated Cell Culture Plates, Corning®
Microplates Cell Culture Plates
Falcon® Cell Culture Plates with a flat bottom and lid
For use when cell attachment is not required such as suspension culture.
Made from crystal grade polystyrene. Sterile and nonpyrogenic.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty