Empty Cassettes and Combs, Electrophoresis, Mini, Snap Gels™, Jule
15C10 P15
66025-449PK 38.24 USD InStock
66025-449 66025-445
Empty Cassettes and Combs, Electrophoresis, Mini, Snap Gels™, Jule
Electrophoresis Systems Vertical Gel Electrophoresis Systems Combs for Vertical Gel Electrophoresis Systems
Empty Snap Gels™ Cassettes are perfect for pouring your own polyacrylamide gels.
  • Save time and money
  • Sealed on three sides so they do not leak
  • Cassettes fit most popular apparatuses
Empty Snap Gels™ Cassettes provides the freedom to make any type of gel formulation. They save time and money typically spent washing and assembling glass plates, which can leak. The Empty Cassettes are sealed on three sides so they do not leak. After gel polymerization, snap off the bottom tab of the cassette to expose the gel to the lower buffer. After electrophoresis, simply use a screwdriver to pry the two cassette halves apart to access the gel. The special plastic cassette material does not inhibit polymerization of acrylamide. A decal is provided for marking the location of sample wells. Cassettes are available in either 0.75 mm, 1.0 mm, or 1.5 mm gel thickness. These cassettes fit most popular apparatuses (see specifications). Form sample wells with the reusable 10 or 15 teeth combs.
Empty Snap Gels™ Cassettes fit the following apparatuses: Hoefer-GE: Mighty Smalls SE250, SE260, MiniVE SE300. Bio-Rad: Protean II, III (For Protean III please request free Jule gaskets), Tetra (and Tetra Transblot), Mini Protean 3 Dodeca. Invitrogen-Novex: Surelock, XCell I and II. Thermo: EC-120. CBS Scientific: MGV-402, MGV-302 (one gel). Owl Scientific: Penguin P8DS, Wolverine P82 (needs adapter), P81 Puffin (single sided) and Fisher Biotech: FB-VE10-1, FB-VE121 (needs adapter).
Plastic Combs are used to form sample wells. Insert the Comb into the cassettes after filling the Cassettes with Gel solution. The Combs are designed to fit perfectly into Jule Empty Snap Gel Cassettes. Remove the Comb after polymerization. The Combs are made of a special plastic material that does not inhibit polymerization. They are available with 10 or 15 teeth and in 0.75 mm, 1.0 mm, or 1.5 mm thickness. Choose the comb thickness to match the Cassette/gel thickness. The combs are reusable.
Ordering information: Empty Snap Gel™ Cassettes are sold in packs of 25. The combs are sold in packs of 15. Power settings and gel pouring instructions are included.