Psychrometer with InfraRed Thermometer, Extech
Supplier: Extech
HD500 HD500-NIST
97014-496EA 627.9 USD
97014-496 10171-748
Psychrometer with InfraRed Thermometer, Extech
Hygrometers Psychrometers
Humidity, Type K Thermometer and 30:1 InfraRed Thermometer
Features a built-in InfraRed Thermometer for non-contact temperature measurement. It sports a large triple LCD backlit display and a built-in USB port. It also features Min/Max, Data hold and Auto power off functions.
Certifications: CE marked.
Ordering information: Three-year warranty. Complete with Windows® compatible software with USB cable, Type K bead wire temperature probe, built-in stand, hard carrying case, and 9V battery