Type J/K Dual Input Thermometer, Extech
Supplier: Extech
TM300 TM300-NIST
89370-160EA 311.42 USD
89370-160 10171-800
Type J/K Dual Input Thermometer, Extech
Thermometers Digital Thermometers General Purpose Digital Thermometers
Dual Input Compact-size Thermometer with Side-Button Controls For Convenient One-hand Operation
Features a large backlit LCD display, readout in °F, °C, or K. Functions include Data Hold, Max/Min/Avg, Offset, and Auto Power off.
Certifications: CE marked.
Ordering information: One year warranty. Complete with two Type K bead wire temperature probes, three AAA batteries, and case.