Pocket Colorimeter™ II, Fluoride (SPADNS II - Arsenic-Free), Hach
2513100
89231-412EA 690.06 USD
89231-412
Pocket Colorimeter™ II, Fluoride (SPADNS II - Arsenic-Free), Hach
The new Pocket Colorimeter™ II Filter Photometer is a true go-anywhere instrument.
  • Single Parameter go anywhere portable photometer
  • USEPA approved for drinking water and wastewater
  • FactoryProgrammed
  • Waterproof, Lightweight, Rugged
  • Backlit LCD Display, Data Logging, Recall
It's lightweight and battery operated, suitable for extended field work or quick, on-the-spot process monitoring. The instrument has two channels in which measurements can be made. Each channel will accept a user-defined calibration curve. Up to 10 standards can be used to determine the calibration. The curve is generated by a point-to-point straight line segment between each standard used. Linear and non-linear positive or negative slope calibrations can be performed. A calibration curve may also be manually entered from the keypad if a previously determined curve has been made on the Colorimeter. At least two data pairs (concentration and absorbance) are required.
Ordering information: (4) AAA batteries required. 2 yr warranty.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.