Would you like to log in again?
Fluoride Standard Solution, 10.0 mg/L as F (NIST), 500 mL, Hach
Supplier: Hach
35949
97021-494EA 44.67 USD InStock
CA359-49 97021-494
Fluoride Standard Solution, 10.0 mg/L as F (NIST), 500 mL, Hach
Standards Environmental Analysis Standards
For accuracy checks in fluoride determinations and calibration of fluoride ion selective electrodes.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty