pH Electrode Storage Solution Powder Pillows, Hach
Supplier: Hach
2657364 2657364
97021-756PK 47.13 USD InStock
97021-756 CA11016-640
pH Electrode Storage Solution Powder Pillows, Hach
Environmental Test Kits
pH Electrode Storage Solution Powder Pillows
Each pillow makes 25 mL of storage solution.
Packaging: Pack of 20 Powder Pillows.