Type J/K Single Input Thermometer, Extech
Supplier: Extech
TM100-NIST TM100
10171-798EA 305.52 USD
10171-798 89370-158
Type J/K Single Input Thermometer, Extech
Thermometers Digital Thermometers General Purpose Digital Thermometers
Compact-size Thermometer with Side-Button Controls For Convenient One-hand Operation
Features a large backlit LCD display, readout in °F, °C, or K. Functions include Data Hold, Max/Min/Avg, Offset, and Auto Power off.
Certifications: CE marked.
Ordering information: One-year warranty. Complete with general purpose Type K bead wire temperature probe and 9V battery.