Would you like to log in again?
MolyVer® 3 Reagent Powder Pillows, 25 mL, Hach
Supplier: Hach
14178-69 1417869
97026-354PK 62.62 USD InStock
CA14178-69 97026-354
MolyVer® 3 Reagent Powder Pillows, 25 mL, Hach
Environmental Test Kits
For determination of high range molybdenum by the mercaptoacetic acid method (Hach method 8036)
Range: up to 40.0 mg/L Mo Sample Size: 25 mL.
Ordering information: Pack of 100 powder pillows.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty