Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Hardness, Iron, and pH Test Kit, Model HA-62B, Hach
Supplier: Hach
183702 183702
89231-250EA 254.67 USD InStock
89231-250 CA11024-946
Hardness, Iron, and pH Test Kit, Model HA-62B, Hach
Environmental Test Kits
Includes color disc and color comparator for colorimetric comparison of iron and pH, as well as a drop-count titrator for hardness determination.
  • Combines frequently measured parameters in one rugged kit
  • Uses color disc and drop count titration methods
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Test kit produces a wide range of pH tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty