NitriVer 3 TNT Reagent Set, Nitrogen-Nitrite, Low Range, Hach
Supplier: Hach
2608345 2608345
97026-522EA 127.26 USD
97026-522 CA97026-522
NitriVer 3 TNT Reagent Set, Nitrogen-Nitrite, Low Range, Hach
Environmental Test Kits
For determination of Nitrite Nitrogen by the NitriVer® 3 Diazotization Test 'N Tube™ Method
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Hach Method 10019. Range: 0.002 - 0.500 mg/L NO2-N.
Ordering information: 50 tests per package.