NitriVer® 3 Nitrite Reagent Powder Pillows, 25 mL, Hach
Supplier: Hach
1406528 14065-99 1406599
10765-226PK 463.08 USD
10765-226 CA14065-99 97026-586
NitriVer® 3 Nitrite Reagent Powder Pillows, 25 mL, Hach
Environmental Test Kits
For low range nitrite determination by the NitriVer® 3 Diazotization Method: EPA Compliant - Hach Method 8507
Range: up to 0.300 mg/L NO2-N Sample Size: 25 mL.
Ordering information: Pack of 100 powder pillows.